DCJTB 98%

BLDPBD19196-250MGEA 146 EUR
BLDPBD19196-250MG BLDPBD19196-100MG BLDPBD19196-1G
DCJTB 98%
DCJTB

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₀H₃₅N₃O
Molecular Weight: 453,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044893
CAS-Nummer: 200052-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR