D(+)-Galactosamin Hydrochlorid 98%

BLDPBD18919-1GEA 6 EUR
BLDPBD18919-1G BLDPBD18919-500G BLDPBD18919-100G BLDPBD18919-10G BLDPBD18919-5G BLDPBD18919-25G
D(+)-Galactosamin Hydrochlorid 98%
D(+)-Galactosamin Hydrochlorid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₁₄ClNO₅
Molecular Weight: 215,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135830
CAS-Nummer: 1772-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR