(S)-(-)-N-Benzyl-α-methylbenzylamin 97%

BLDPBD18889-25GEA 18 EUR
BLDPBD18889-25G BLDPBD18889-500G BLDPBD18889-1G BLDPBD18889-10G BLDPBD18889-100G BLDPBD18889-5G
(S)-(-)-N-Benzyl-α-methylbenzylamin 97%
(S)-(-)-N-Benzyl-α-methylbenzylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₁₇N
Molecular Weight: 211,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066325
CAS-Nummer: 17480-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR