(3-Methylphenyl)methansulfonsäurechlorid 95%

BLDPBD188634-1GEA 77 EUR
BLDPBD188634-1G BLDPBD188634-250MG
(3-Methylphenyl)methansulfonsäurechlorid 95%
(3-Methylphenyl)methanesulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₉ClO₂S
Molecular Weight: 204,6700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04117457
CAS-Nummer: 53531-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR