4-Iodbiphenyl 97%

Synonyme: 1-Jod-4-phenylbenzol1-Iod-4-phenylbenzol

BLDPBD18647-1000GEA 433 EUR
BLDPBD18647-1000G BLDPBD18647-10G BLDPBD18647-100G BLDPBD18647-0.025G BLDPBD18647-25G BLDPBD18647-500G BLDPBD18647-5G
4-Iodbiphenyl 97%
4-Iodbiphenyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₉I
Molecular Weight: 280,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019028
CAS-Nummer: 1591-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR