4,7-Dibrom-2,1,3-benzothiadiazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD18514-1GEA 7 EUR
BLDPBD18514-1G BLDPBD18514-10G BLDPBD18514-25G BLDPBD18514-5G
4,7-Dibrom-2,1,3-benzothiadiazol 98%
4,7-Dibromo-2,1,3-benzothiadiazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₂Br₂N₂S
Molecular Weight: 293,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00658844
CAS-Nummer: 15155-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR