N(α)-Cbz-L-histidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD18488-5GEA 33 EUR
BLDPBD18488-5G BLDPBD18488-1G BLDPBD18488-25G
N(α)-Cbz-L-histidin 98%
N(α)-Cbz-L-histidin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₁₅N₃O₄
Molecular Weight: 289,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065960
CAS-Nummer: 14997-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR