4-Cyanpyridin-N-oxid 98%

BLDPBD18467-10GEA 13 EUR
BLDPBD18467-10G BLDPBD18467-5G BLDPBD18467-100G BLDPBD18467-25G
4-Cyanpyridin-N-oxid 98%
4-Cyanpyridin-N-oxid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄N₂O
Molecular Weight: 120,1088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006205
CAS-Nummer: 14906-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR