3-Iod-1H-pyrazol 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD182449-10GEA
103
EUR
BLDPBD182449-10G
BLDPBD182449-5G
BLDPBD182449-1G
BLDPBD182449-25G
BLDPBD182449-0.025
BLDPBD182449-100G
3-Iod-1H-pyrazol 98%
3-Iodo-1H-pyrazole
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₃H₃IN₂ Molecular Weight: 193,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12022324 CAS-Nummer: 4522-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...