α,3-Dibrom-2-fluortoluol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD182218-10GEA 166 EUR
BLDPBD182218-10G BLDPBD182218-1G BLDPBD182218-25G BLDPBD182218-250MG BLDPBD182218-5G
α,3-Dibrom-2-fluortoluol 97%
α,3-Dibrom-2-fluortoluol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅Br₂F
Molecular Weight: 267,92 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09842442
CAS-Nummer: 149947-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR