Methyl-2-iod-5-methylbenzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD182123-25GEA 168 EUR
BLDPBD182123-25G BLDPBD182123-5G BLDPBD182123-1G
Methyl-2-iod-5-methylbenzoat 98%
Methyl-2-iod-5-methylbenzoat

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₉IO₂
Molecular Weight: 276,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12198129
CAS-Nummer: 103440-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR