Ethyl-2-aminonicotinat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD18079-25GEA 97 EUR
BLDPBD18079-25G BLDPBD18079-100G BLDPBD18079-10G BLDPBD18079-5G BLDPBD18079-1G
Ethyl-2-aminonicotinat 97%
Ethyl-2-aminonicotinat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569452
CAS-Nummer: 13362-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR