4-Hydroxydiphenylamin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD17817-1GEA 7 EUR
BLDPBD17817-1G
4-Hydroxydiphenylamin 98%
4-Hydroxydiphenylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₁NO
Molecular Weight: 185,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020142
CAS-Nummer: 122-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR