2,5-Difluor-4-nitrobenzoesäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD17666-10GEA 53 EUR
BLDPBD17666-10G BLDPBD17666-100G BLDPBD17666-500G BLDPBD17666-5G BLDPBD17666-250MG BLDPBD17666-1G BLDPBD17666-25G
2,5-Difluor-4-nitrobenzoesäure 95%
2,5-Difluoro-4-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃F₂NO₄
Molecular Weight: 203,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425705
CAS-Nummer: 116465-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR