1-(4-Chlorphenyl)cyclopropanamin Hydrochlorid 98%
|
|
BLDPBD173975-100MGEA
34
EUR
BLDPBD173975-100MG
BLDPBD173975-250MG
BLDPBD173975-1G
1-(4-Chlorphenyl)cyclopropanamin Hydrochlorid 98%
1-(4-Chlorphenyl)cyclopropanamin Hydrochlorid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₉H₁₁Cl₂N Molecular Weight: 204,0963 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07995727 CAS-Nummer: 1009102-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...