1,3-Benzothiazol-6-ylboronic acid 95%

BLDPBD173576-250MGEA 87 EUR
BLDPBD173576-250MG BLDPBD173576-100MG BLDPBD173576-5G BLDPBD173576-1G
1,3-Benzothiazol-6-ylboronic acid 95%
1,3-Benzothiazol-6-ylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BNO₂S
Molecular Weight: 179,004 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05664651
CAS-Nummer: 499769-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR