PIPES Sesquinatriumsalz 98%

Synonyme: Piperazin-N,N'-bis-2-ethansulfonsäure SesquinatriumsalzPiperazin-1,4-bis(2-ethansulfonsäure) Sesquinatriumsalz2,2'-(Piperazin-1,4-diyl)bis(ethansulfonsäure) SesquinatriumsalzPIPES-Na1,5Sesquinatriumpiperazin-1,4-diethansulfonat

BLDPBD17230-5GEA 6 EUR
BLDPBD17230-5G
PIPES Sesquinatriumsalz 98%
PIPES Sesquinatriumsalz
Formel: C₁₆H₃₃N₄Na₃O₁₂S₄
Molecular Weight: 670,69 g/mol
MDL: MFCD00066393
CAS-Nummer: 100037-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR