3-Brom-N-methylbenzamid 98%

BLDPBD172263-10GEA 149 EUR
BLDPBD172263-10G BLDPBD172263-5G BLDPBD172263-25G BLDPBD172263-250MG BLDPBD172263-1G
3-Brom-N-methylbenzamid 98%
3-Brom-N-methylbenzamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈BrNO
Molecular Weight: 214,0592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01060879
CAS-Nummer: 49834-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR