3,4-Dihydroxybenzophenon 98%
|
|
BLDPBD17062-5GEA
14
EUR
BLDPBD17062-5G
BLDPBD17062-100G
BLDPBD17062-25G
BLDPBD17062-10G
3,4-Dihydroxybenzophenon 98%
3,4-Dihydroxybenzophenon
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214,2200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00477203 CAS-Nummer: 10425-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...