1-Naphthylacetamid 97%
|
|
BLDPBD17057-10GEA
11
EUR
BLDPBD17057-0.025G
BLDPBD17057-10G
BLDPBD17057-25G
1-Naphthylacetamid 97%
1-Naphthylacetamid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₂H₁₁NO Molecular Weight: 185,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004047 CAS-Nummer: 86-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...