4-(N,N-Diphenylamino)benzaldehyd 96%

BLDPBD16988-1GEA 7 EUR
BLDPBD16988-1G BLDPBD16988-100G BLDPBD16988-500G BLDPBD16988-5G BLDPBD16988-25G BLDPBD16988-10G
4-(N,N-Diphenylamino)benzaldehyd 96%
4-(N,N-Diphenylamino)benzaldehyd

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₉H₁₅NO
Molecular Weight: 273,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145131
CAS-Nummer: 4181-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR