4,5-Difluor-2-nitrotoluol 98%

BLDPBD168149-1GEA 7 EUR
BLDPBD168149-1G BLDPBD168149-5G BLDPBD168149-10G BLDPBD168149-25G BLDPBD168149-100G
4,5-Difluor-2-nitrotoluol 98%
4,5-Difluor-2-nitrotoluol

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₅F₂NO₂
Molecular Weight: 173,12 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04973772
CAS-Nummer: 127371-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR