1-Methyl-4-indolboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD168087-1GEA 117 EUR
BLDPBD168087-1G BLDPBD168087-5G BLDPBD168087-250MG
1-Methyl-4-indolboronsäurepinakolester 98%
1-Methyl-4-indolboronsäurepinakolester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257,1400 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08690266
CAS-Nummer: 898289-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR