Methyl-3-iod-1H-indazol-6-carboxylat 95%

BLDPBD168069-25GEA 478 EUR
BLDPBD168069-25G BLDPBD168069-250MG BLDPBD168069-5G BLDPBD168069-10G BLDPBD168069-1G
Methyl-3-iod-1H-indazol-6-carboxylat 95%
Methyl-3-iod-1H-indazol-6-carboxylat

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₇IN₂O₂
Molecular Weight: 302,0686 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781616
CAS-Nummer: 885518-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR