4-Chloroquinoline-3-carbaldehyde 95%

BLDPBD166898-100MGEA 132 EUR
BLDPBD166898-100MG BLDPBD166898-250MG BLDPBD166898-1G
4-Chloroquinoline-3-carbaldehyde 95%
4-Chloroquinoline-3-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₆ClNO
Molecular Weight: 191,614 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03787717
CAS-Nummer: 201420-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR