Senicapoc 98%

BLDPBD165423-100MGEA 117 EUR
BLDPBD165423-100MG BLDPBD165423-1G BLDPBD165423-250MG
Senicapoc 98%
Senicapoc

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₀H₁₅F₂NO
Molecular Weight: 323,34 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09027349
CAS-Nummer: 289656-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR