5-Fluor-2-formylphenylboronsäure 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD165409-250MGEA
7
EUR
BLDPBD165409-250MG
BLDPBD165409-10G
BLDPBD165409-100G
BLDPBD165409-5G
BLDPBD165409-25G
BLDPBD165409-1G
5-Fluor-2-formylphenylboronsäure 98%
5-Fluor-2-formylphenylboronsäure
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₆BFO₃ Molecular Weight: 167,9300 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09953731 CAS-Nummer: 1256355-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...