Methyl 5-chloro-2-methylbenzoate 97%

BD165299-10G BD165299-1G BD165299-25G BD165299-5G
BLDPBD165299-10GEA 118 EUR
BLDPBD165299-10G BLDPBD165299-1G BLDPBD165299-25G BLDPBD165299-5G
Methyl 5-chloro-2-methylbenzoate 97%
Methyl 5-chloro-2-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Ambient
MDL: MFCD09831993
CAS-Nummer: 99585-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR