6-Iodobenzo[d]thiazole 97%

BD165242-100MG BD165242-250MG BD165242-1G
BLDPBD165242-100MGEA 62 EUR
BLDPBD165242-100MG BLDPBD165242-250MG BLDPBD165242-1G
6-Iodobenzo[d]thiazole 97%
6-Iodobenzo[d]thiazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₄INS
Molecular Weight: 261,08 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD06659655
CAS-Nummer: 654070-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR