2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol 97%

BD165241-500G BD165241-25G BD165241-10G BD165241-100G
BLDPBD165241-500GEA 405 EUR
BLDPBD165241-500G BLDPBD165241-25G BLDPBD165241-10G BLDPBD165241-100G
2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol 97%
2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₃H₂₉Cl₂NO
Molecular Weight: 406,39 g/mol
Lagertemperatur: Ambient
MDL: MFCD12923306
CAS-Nummer: 69759-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR