2-Brom-5-iodphenol 97%

BLDPBD165177-5GEA 292 EUR
BLDPBD165177-5G BLDPBD165177-1G BLDPBD165177-25G BLDPBD165177-100MG BLDPBD165177-10G BLDPBD165177-250MG
2-Brom-5-iodphenol 97%
2-Brom-5-iodphenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄BrIO
Molecular Weight: 298,9000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907878
CAS-Nummer: 932372-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR