3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 98%

BLDPBD165169-1GEA 14 EUR
BLDPBD165169-1G BLDPBD165169-100G BLDPBD165169-25G BLDPBD165169-10G BLDPBD165169-5G
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 98%
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₃ClF₃NO
Molecular Weight: 197,541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153095
CAS-Nummer: 76041-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR