3,5-Bis(tert-butyl)-2-methoxybenzaldehyde 97%

Synonyme: 3,5-Di-tert-butyl-2-methoxybenzaldehyde

BD16503-1G BD16503-5G BD16503-25G BD16503-250MG
BLDPBD16503-1GEA 47 EUR
BLDPBD16503-1G BLDPBD16503-5G BLDPBD16503-25G BLDPBD16503-250MG
3,5-Bis(tert-butyl)-2-methoxybenzaldehyde 97%
3,5-Bis(tert-butyl)-2-methoxybenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₆H₂₄O₂
Molecular Weight: 248,361 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD02093684
CAS-Nummer: 135546-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR