7-Bromo-4-fluoroindoline-2,3-dione 95%

BD164975-1G BD164975-250MG BD164975-100MG
BLDPBD164975-1GEA 380 EUR
BLDPBD164975-1G BLDPBD164975-250MG BLDPBD164975-100MG
7-Bromo-4-fluoroindoline-2,3-dione 95%
7-Bromo-4-fluoroindoline-2,3-dione

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₃BrFNO₂
Molecular Weight: 244,01 g/mol
Lagertemperatur: Ambient
MDL: MFCD17677300
CAS-Nummer: 1245648-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR