(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid 97%

BLDPBD16493-5GEA 37 EUR
BLDPBD16493-5G BLDPBD16493-10G BLDPBD16493-1G BLDPBD16493-250MG BLDPBD16493-25G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₃NO₅
Molecular Weight: 309,3575 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00065601
CAS-Nummer: 76757-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR