3-(Trifluormethyl)-5-pyridinboronsäure 95%

Synonyme: (5-(Trifluoromethyl)pyridin-3-yl)boronic acid,(5-Trifluoromethylpyridin-3-yl)boronic acid

BD164862-100MG
BLDPBD164862-100MGEA 46 EUR
BLDPBD164862-100MG
3-(Trifluormethyl)-5-pyridinboronsäure 95%
3-(Trifluormethyl)-5-pyridinboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅BF₃NO₂
Molecular Weight: 190,9156 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09952041
CAS-Nummer: 947533-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR