2-Methoxy-6-methylpyridin-4-amine 95%

BD164836-250MG BD164836-100MG BD164836-5G BD164836-50MG BD164836-1G
BLDPBD164836-250MGEA 59 EUR
BLDPBD164836-250MG BLDPBD164836-100MG BLDPBD164836-5G BLDPBD164836-50MG BLDPBD164836-1G
2-Methoxy-6-methylpyridin-4-amine 95%
2-Methoxy-6-methylpyridin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Ambient
MDL: MFCD15529703
CAS-Nummer: 89943-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR