5-Bromo-2-propoxypyridine 98%

BD164795-1G BD164795-5G BD164795-250MG
BLDPBD164795-1GEA 102 EUR
BLDPBD164795-1G BLDPBD164795-5G BLDPBD164795-250MG
5-Bromo-2-propoxypyridine 98%
5-Bromo-2-propoxypyridine

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₀BrNO
Molecular Weight: 216,07 g/mol
Lagertemperatur: Ambient
MDL: MFCD09743641
CAS-Nummer: 850142-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR