1-Boc-4-(4-nitrobenzyl)piperazine 98%

BLDPBD16478-250MGEA 19 EUR
BLDPBD16478-250MG BLDPBD16478-1G BLDPBD16478-5G
1-Boc-4-(4-nitrobenzyl)piperazine 98%
1-Boc-4-(4-nitrobenzyl)piperazine

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₆H₂₃N₃O₄
Molecular Weight: 321,3700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03407491
CAS-Nummer: 130636-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR