tert-Butyl (3-methoxyphenyl)carbamate 98%

BD164739-100G BD164739-5G BD164739-25G
BLDPBD164739-100GEA 206 EUR
BLDPBD164739-100G BLDPBD164739-5G BLDPBD164739-25G
tert-Butyl (3-methoxyphenyl)carbamate 98%
tert-Butyl (3-methoxyphenyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₇NO₃
Molecular Weight: 223,26 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07127720
CAS-Nummer: 60144-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR