Ethyl-1-acetylcyclopropane-1-carboxylate 97%

BLDPBD164683-25GEA 416 EUR
BLDPBD164683-25G BLDPBD164683-250MG BLDPBD164683-5G BLDPBD164683-1G BLDPBD164683-100G
Ethyl-1-acetylcyclopropane-1-carboxylate 97%
Ethyl-1-acetylcyclopropane-1-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₂O₃
Molecular Weight: 156,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00480999
CAS-Nummer: 32933-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR