(S)-5-Hydroxypiperidin-2-one 98%

BLDPBD164656-5GEA 648 EUR
BLDPBD164656-5G BLDPBD164656-1G BLDPBD164656-250MG BLDPBD164656-100MG
(S)-5-Hydroxypiperidin-2-one 98%
(S)-5-Hydroxypiperidin-2-one

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06202374
CAS-Nummer: 24211-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR