Boc-D-aspartic acid β-benzyl ester 97%

Synonyme: N-Boc-D-aspartic acid 4-benzyl ester,Boc-D-Asp(OBzl)-OH

BD16462-10G BD16462-1G BD16462-500G BD16462-25G BD16462-250MG BD16462-100G BD16462-5G
BLDPBD16462-10GEA 26 EUR
BLDPBD16462-10G BLDPBD16462-1G BLDPBD16462-500G BLDPBD16462-25G BLDPBD16462-250MG BLDPBD16462-100G BLDPBD16462-5G
Boc-D-aspartic acid β-benzyl ester 97%
Boc-D-aspartic acid beta-benzyl ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₆H₂₁NO₆
Molecular Weight: 323,345 g/mol
Lagertemperatur: Ambient
MDL: MFCD00038255
CAS-Nummer: 51186-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR