6-Fluoro-3-methyl-1H-indazole 95%

BD164617-25G BD164617-250MG BD164617-1G BD164617-100MG BD164617-5G
BLDPBD164617-25GEA 2921 EUR
BLDPBD164617-25G BLDPBD164617-250MG BLDPBD164617-1G BLDPBD164617-100MG BLDPBD164617-5G
6-Fluoro-3-methyl-1H-indazole 95%
6-Fluoro-3-methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇FN₂
Molecular Weight: 150,15 g/mol
Lagertemperatur: Ambient
MDL: MFCD08692363
CAS-Nummer: 159305-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR