4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%

Synonyme: Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enecarboxylate

BD164599-250MG BD164599-100MG BD164599-1G BD164599-5G
BLDPBD164599-250MGEA 135 EUR
BLDPBD164599-250MG BLDPBD164599-100MG BLDPBD164599-1G BLDPBD164599-5G
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₄H₂₃BO₄
Molecular Weight: 266,141 g/mol
Lagertemperatur: Freezer
MDL: MFCD11520532
CAS-Nummer: 151075-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR