Ethylimidazo[1,2-a]pyridin-3-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD164573-1GEA 18 EUR
BLDPBD164573-1G BLDPBD164573-5G BLDPBD164573-25G
Ethylimidazo[1,2-a]pyridin-3-carboxylat 97%
Ethylimidazo[1,2-a]pyridin-3-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656766
CAS-Nummer: 123531-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR