(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 97%

BD164571-5G BD164571-100MG BD164571-250MG BD164571-1G
BLDPBD164571-5GEA 983 EUR
BLDPBD164571-5G BLDPBD164571-100MG BLDPBD164571-250MG BLDPBD164571-1G
(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 97%
(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₈N₂O₃
Molecular Weight: 214,26 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD13195282
CAS-Nummer: 172913-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR