2-Chloro-6-cyclopropylpyrazine 95%

BLDPBD164561-100MGEA 118 EUR
BLDPBD164561-100MG BLDPBD164561-1G BLDPBD164561-250MG
2-Chloro-6-cyclopropylpyrazine 95%
2-Chloro-6-cyclopropylpyrazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇ClN₂
Molecular Weight: 154,59 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14702732
CAS-Nummer: 1209457-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR