3-Bromo-1-methyl-1H-indazol-6-amine 95%

BLDPBD164557-1GEA 192 EUR
BLDPBD164557-1G BLDPBD164557-250MG BLDPBD164557-100MG
3-Bromo-1-methyl-1H-indazol-6-amine 95%
3-Bromo-1-methyl-1H-indazol-6-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₈BrN₃
Molecular Weight: 226,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12827698
CAS-Nummer: 1203181-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR