(E)-But-2-ene-1,4-diamine dihydrochloride 95%

BD164554-1G BD164554-250MG BD164554-5G
BLDPBD164554-1GEA 146 EUR
BLDPBD164554-1G BLDPBD164554-250MG BLDPBD164554-5G
(E)-But-2-ene-1,4-diamine dihydrochloride 95%
(E)-But-2-ene-1,4-diamine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₄H₁₂Cl₂N₂
Molecular Weight: 159,05 g/mol
Lagertemperatur: Ambient
MDL: MFCD16039313
CAS-Nummer: 119874-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR